Product Details

SNP ID
rs1870874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:40110654 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATGCTATCATTATTTTCAAGATT[A/C]TTATTAAAACAAGCTGAAACAACAA
Phenotype
MIM: 608817
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LRRC4C PubMed Links

Gene Details

Gene
LRRC4C
Gene Name
leucine rich repeat containing 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258419.1 6863 Intron NP_001245348.1
NM_020929.2 6863 Intron NP_065980.1
XM_011520238.2 6863 UTR 3 XP_011518540.1
XM_011520239.2 6863 UTR 3 XP_011518541.1
XM_011520240.2 6863 UTR 3 XP_011518542.1
XM_011520241.2 6863 UTR 3 XP_011518543.1
XM_011520242.2 6863 UTR 3 XP_011518544.1
XM_011520243.2 6863 UTR 3 XP_011518545.1
XM_011520244.2 6863 UTR 3 XP_011518546.1
XM_017018070.1 6863 UTR 3 XP_016873559.1
XM_017018071.1 6863 UTR 3 XP_016873560.1
XM_017018072.1 6863 UTR 3 XP_016873561.1
XM_017018073.1 6863 UTR 3 XP_016873562.1
XM_017018074.1 6863 UTR 3 XP_016873563.1
XM_017018075.1 6863 UTR 3 XP_016873564.1
XM_017018076.1 6863 UTR 3 XP_016873565.1
XM_017018077.1 6863 UTR 3 XP_016873566.1
XM_017018078.1 6863 UTR 3 XP_016873567.1
XM_017018079.1 6863 UTR 3 XP_016873568.1

View Full Product Details