Product Details

SNP ID
rs14995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180790619 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCTTTCATGAAACTGAGGTCAGG[A/C]AAGAAACAAAAATCCACCAAGTCCT
Phenotype
MIM: 160995
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MGAT1 PubMed Links

Gene Details

Gene
MGAT1
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114617.1 3119 UTR 3 NP_001108089.1
NM_001114618.1 3119 UTR 3 NP_001108090.1
NM_001114619.1 3119 UTR 3 NP_001108091.1
NM_001114620.1 3119 UTR 3 NP_001108092.1
NM_002406.3 3119 UTR 3 NP_002397.2
XM_005265915.1 3119 UTR 3 XP_005265972.1
XM_005265916.1 3119 UTR 3 XP_005265973.1
XM_006714866.1 3119 UTR 3 XP_006714929.1
XM_011534559.1 3119 UTR 3 XP_011532861.1
XM_011534560.1 3119 UTR 3 XP_011532862.1
XM_011534561.1 3119 UTR 3 XP_011532863.1
XM_011534562.1 3119 UTR 3 XP_011532864.1
XM_011534563.2 3119 UTR 3 XP_011532865.1
XM_017009486.1 3119 UTR 3 XP_016864975.1
XM_017009487.1 3119 UTR 3 XP_016864976.1

View Full Product Details