Product Details

SNP ID
rs1065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:132403522 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATACCCAATGAAGCAGGCTATGGA[A/T]TAAAAGAAACAACAAATGGATTCAT
Phenotype
MIM: 600632
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OPCML PubMed Links

Gene Details

Gene
OPCML
Gene Name
opioid binding protein/cell adhesion molecule like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012393.2 18512 Intron NP_001012393.1
NM_001319103.1 18512 Intron NP_001306032.1
NM_001319104.1 18512 Intron NP_001306033.1
NM_001319105.1 18512 Intron NP_001306034.1
NM_001319106.1 18512 Intron NP_001306035.1
NM_002545.4 18512 Intron NP_002536.1
XM_006718846.2 18512 UTR 3 XP_006718909.1
XM_011542856.2 18512 UTR 3 XP_011541158.1

View Full Product Details