Product Details
- SNP ID
-
rs12801553
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:73882345 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTTTTTTTGAAGGTAACTTGTAG[A/G]TTCAGTTTTCTTTATGAATTGGAGT
- Phenotype
-
MIM: 608016
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COA4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111853342] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COA4
- Gene Name
- cytochrome c oxidase assembly factor 4 homolog
There are no transcripts associated with this gene.
- Gene
- PAAF1
- Gene Name
- proteasomal ATPase associated factor 1
View Full Product Details