Product Details

SNP ID
rs10899392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77591333 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCTCAGATGCAGTGGGCAATCTA[C/G]TCCTAAGTGGGAGGAGGAAATCCAA
Phenotype
MIM: 609914
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AQP11 PubMed Links

Gene Details

Gene
AQP11
Gene Name
aquaporin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173039.2 Intron NP_766627.1
XM_005273917.4 Intron XP_005273974.1
Gene
LOC107987163
Gene Name
uncharacterized LOC107987163
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017018644.1 Intron XP_016874133.1

View Full Product Details