Product Details

SNP ID
rs71464099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73951558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCAGCATTGACTAAACTACATT[C/G]CTGAATACTTCCATCGGATCTTAAG
Phenotype
MIM: 610263
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB13 PubMed Links

Gene Details

Gene
DNAJB13
Gene Name
DnaJ heat shock protein family (Hsp40) member B13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153614.3 Intron NP_705842.2
XM_005273984.3 Intron XP_005274041.1
XM_011545004.2 Intron XP_011543306.1
XM_011545005.2 Intron XP_011543307.1
XM_011545007.2 Intron XP_011543309.1
XM_011545009.2 Intron XP_011543311.1
XM_011545013.2 Intron XP_011543315.1
XM_011545014.2 Intron XP_011543316.1
XM_011545015.2 Intron XP_011543317.1
XM_017017675.1 Intron XP_016873164.1
XM_017017676.1 Intron XP_016873165.1
XM_017017677.1 Intron XP_016873166.1
XM_017017678.1 Intron XP_016873167.1
XM_017017679.1 Intron XP_016873168.1

View Full Product Details