Product Details

SNP ID
rs1916743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:167307220 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACGAGTTAATTTGAAGTGAAATA[A/G]GAAGAAACAGTGTGAAGTATATATG
Phenotype
MIM: 603093
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B3GALT1 PubMed Links

Gene Details

Gene
B3GALT1
Gene Name
beta-1,3-galactosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020981.3 Intron NP_066191.1
XM_005246931.3 Intron XP_005246988.1
XM_006712819.3 Intron XP_006712882.1
XM_011512085.2 Intron XP_011510387.1

View Full Product Details