Product Details

SNP ID
rs2042772
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:159805547 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AACACCAATTTGGGAACATCACTAC[C/T]TGTATATTATCCTACTTTGATAGCA
Phenotype
MIM: 612246 MIM: 604524
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD302 PubMed Links

Gene Details

Gene
CD302
Gene Name
CD302 molecule
There are no transcripts associated with this gene.

Gene
LY75
Gene Name
lymphocyte antigen 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002349.3 Intron NP_002340.2
Gene
LY75-CD302
Gene Name
LY75-CD302 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198759.1 Intron NP_001185688.1
NM_001198760.1 Intron NP_001185689.1

View Full Product Details