Product Details

SNP ID
rs2008493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1561410 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACTTCTGCCCGCCACCTCCATGA[C/T]CCCTGGACTCCCCGCCAGGGCGGGG
Phenotype
MIM: 602038
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP8 PubMed Links

Gene Details

Gene
DUSP8
Gene Name
dual specificity phosphatase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004420.2 Intron NP_004411.2
XM_011519932.2 Intron XP_011518234.1
XM_011519933.2 Intron XP_011518235.1
Gene
LOC101927562
Gene Name
uncharacterized LOC101927562
There are no transcripts associated with this gene.

View Full Product Details