Product Details

SNP ID
rs1050963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1753042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACTGAACAGGTAGGGTGGCAGA[A/G]CCCAGCTGGGCCCAAGCTGGGCAGA
Phenotype
MIM: 116840
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTSD PubMed Links

Gene Details

Gene
CTSD
Gene Name
cathepsin D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001909.4 1833 UTR 3 NP_001900.1
Gene
IFITM10
Gene Name
interferon induced transmembrane protein 10
There are no transcripts associated with this gene.

View Full Product Details