Product Details

SNP ID
rs10124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56469731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGATCATCTTCCTCATCCCCTA[C/T]CTTGTGAAAGCTAGGCATACAGCCA
Phenotype
MIM: 606365
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLS2 PubMed Links

Gene Details

Gene
GLS2
Gene Name
glutaminase 2
There are no transcripts associated with this gene.

Gene
SPRYD4
Gene Name
SPRY domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207344.3 853 UTR 3 NP_997227.1

View Full Product Details