Product Details

SNP ID
rs1059859
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56638219 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGGAAACCTTAAATACTGTTCA[A/G]AAAGAATATATCTTCAATCAAGGCT
Phenotype
MIM: 102910 MIM: 605682
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ATP5B PubMed Links
Additional Information
For this assay, SNP(s) [rs149735193] are located under a probe and SNP(s) [rs149735193] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ATP5B
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001686.3 1799 UTR 3 NP_001677.2
Gene
BAZ2A
Gene Name
bromodomain adjacent to zinc finger domain 2A
There are no transcripts associated with this gene.

Gene
SNORD59A
Gene Name
small nucleolar RNA, C/D box 59A
There are no transcripts associated with this gene.

Gene
SNORD59B
Gene Name
small nucleolar RNA, C/D box 59B
There are no transcripts associated with this gene.

View Full Product Details