Product Details

SNP ID
rs8315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185502282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGGGTGCGTGCGTGCGTGCCAC[A/G]CCTGCAGAGACTTAAGCTTTGGGAT
Phenotype
MIM: 605889
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDLIM3 PubMed Links

Gene Details

Gene
PDLIM3
Gene Name
PDZ and LIM domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114107.4 1087 UTR 3 NP_001107579.1
NM_001257962.1 1087 UTR 3 NP_001244891.1
NM_001257963.1 1087 UTR 3 NP_001244892.1
NM_014476.5 1087 UTR 3 NP_055291.2

View Full Product Details