Product Details

SNP ID
rs1840680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:157438240 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCCCCTTACACCCGATCCGACTA[A/G]GCGCTGCAGGAGTCCACTGGCCCGT
Phenotype
MIM: 602492 MIM: 609594
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTX3 PubMed Links

Gene Details

Gene
PTX3
Gene Name
pentraxin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002852.3 Intron NP_002843.2
Gene
VEPH1
Gene Name
ventricular zone expressed PH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167911.1 Intron NP_001161383.1
NM_001167912.1 Intron NP_001161384.1
NM_001167915.1 Intron NP_001161387.1
NM_001167916.1 Intron NP_001161388.1
NM_001167917.1 Intron NP_001161389.1
NM_024621.2 Intron NP_078897.2
XM_011513134.2 Intron XP_011511436.1
XM_011513135.1 Intron XP_011511437.1

View Full Product Details