Product Details

SNP ID
rs1947058
Assay Type
Validated
NCBI dbSNP Submissions
33
Location
Chr.1:211478668 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAATGGGACATCCCTGAAATTGGGG[A/T]GAGAACCTAGGTCTTGCCAAAAGGC
Phenotype
MIM: 180040
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RD3 PubMed Links

Gene Details

Gene
RD3
Gene Name
retinal degeneration 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164688.1 1487 UTR 3 NP_001158160.1
NM_183059.2 1487 UTR 3 NP_898882.1
XM_017001151.1 1487 UTR 3 XP_016856640.1

View Full Product Details