Product Details

SNP ID
rs10766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73560872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCCAAAATAGTTTACACCTTCT[A/T]CACTGAAGCATTGTTTAAAATGTAC
Phenotype
MIM: 612820 MIM: 615176
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NPTN PubMed Links

Gene Details

Gene
NPTN
Gene Name
neuroplastin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161363.1 1573 UTR 3 NP_001154835.1
NM_001161364.1 1573 UTR 3 NP_001154836.1
NM_012428.3 1573 UTR 3 NP_036560.1
NM_017455.3 1573 UTR 3 NP_059429.1
XM_011521452.2 1573 Intron XP_011519754.1
Gene
NPTN-IT1
Gene Name
NPTN intronic transcript 1
There are no transcripts associated with this gene.

Gene
REC114
Gene Name
REC114 meiotic recombination protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042367.1 1573 Intron NP_001035826.1
XM_017022083.1 1573 Intron XP_016877572.1

View Full Product Details