Product Details
- SNP ID
-
rs1107215
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
29
- Location
-
Chr.1:212692405 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGACGAAAGAGCAAAAAAAACCC[A/C]AAAAAACCAGAGGCTCAAGTATAGG
- Phenotype
-
MIM: 612470
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BATF3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114195608] are located under a probe and SNP(s) [rs116598640] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BATF3
- Gene Name
- basic leucine zipper ATF-like transcription factor 3
View Full Product Details