Product Details
- SNP ID
-
rs1878672
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
54
- Location
-
Chr.1:206770368 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAACAGGGGAAACCATTAGTAGTGA[C/G]GGCATCATTTCAGTTGGAGAAATAA
- Phenotype
-
MIM: 124092
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IL10
PubMed Links
Gene Details
- Gene
- IL10
- Gene Name
- interleukin 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000572.2 |
|
Intron |
|
|
NP_000563.1 |
View Full Product Details