Product Details

SNP ID
rs1941273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:32944332 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCTATTTAGGGAATCATTAAATA[C/T]ATCAGTGTTCAGAACTCAGAGCATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC178 PubMed Links

Gene Details

Gene
CCDC178
Gene Name
coiled-coil domain containing 178
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105528.1 Intron NP_001098998.1
NM_001308126.1 Intron NP_001295055.1
NM_198995.2 Intron NP_945346.2
XM_011525948.1 Intron XP_011524250.1
XM_011525951.1 Intron XP_011524253.1
XM_011525954.2 Intron XP_011524256.1
XM_011525955.1 Intron XP_011524257.1
XM_017025721.1 Intron XP_016881210.1
XM_017025722.1 Intron XP_016881211.1
XM_017025723.1 Intron XP_016881212.1
XM_017025724.1 Intron XP_016881213.1
XM_017025725.1 Intron XP_016881214.1

View Full Product Details