Product Details

SNP ID
rs7886
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:18128428 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCAGTGAAGACACATATTACATAAA[A/T]GATCAACTACCTAAGGATATCTCAT
Phenotype
MIM: 608072 MIM: 187680
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NHLRC1 PubMed Links

Gene Details

Gene
NHLRC1
Gene Name
NHL repeat containing E3 ubiquitin protein ligase 1
There are no transcripts associated with this gene.

Gene
TPMT
Gene Name
thiopurine S-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000367.3 3449 UTR 3 NP_000358.1
XM_011514839.2 3449 UTR 3 XP_011513141.1
XM_017011241.1 3449 UTR 3 XP_016866730.1

View Full Product Details