Product Details

SNP ID
rs1886330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:18161998 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAATTAAGGGTTAAGGTTCAATAA[A/C]ATCGAGAAAACAAATCCTATTTGTT
Phenotype
MIM: 613081 MIM: 187680
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KDM1B PubMed Links

Gene Details

Gene
KDM1B
Gene Name
lysine demethylase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153042.3 Intron NP_694587.3
XM_005248925.1 Intron XP_005248982.1
XM_005248926.1 Intron XP_005248983.1
XM_005248928.1 Intron XP_005248985.1
XM_011514385.1 Intron XP_011512687.1
XM_011514386.1 Intron XP_011512688.1
XM_017010440.1 Intron XP_016865929.1
XM_017010441.1 Intron XP_016865930.1
XM_017010442.1 Intron XP_016865931.1
XM_017010443.1 Intron XP_016865932.1
XM_017010445.1 Intron XP_016865934.1
XM_017010446.1 Intron XP_016865935.1
Gene
TPMT
Gene Name
thiopurine S-methyltransferase
There are no transcripts associated with this gene.

View Full Product Details