Product Details

SNP ID
rs8057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:43886447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAGCTATTAGATATTTAGAACA[C/T]TCATAAGCCCACCTACAGTATAGTT
Phenotype
MIM: 605460 MIM: 607544
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCG8 PubMed Links

Gene Details

Gene
ABCG8
Gene Name
ATP binding cassette subfamily G member 8
There are no transcripts associated with this gene.

Gene
LRPPRC
Gene Name
leucine rich pentatricopeptide repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133259.3 6396 UTR 3 NP_573566.2
XM_006711915.2 6396 Intron XP_006711978.1
XM_006711916.3 6396 Intron XP_006711979.1
XM_011532473.1 6396 Intron XP_011530775.1
XM_017003117.1 6396 Intron XP_016858606.1

View Full Product Details