Product Details

SNP ID
rs11512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100466570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATTATTATTTTAATAACAACCT[G/T]AATCCCAGAACTTCCAGATACTGTC
Phenotype
MIM: 611914
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C7orf61 PubMed Links

Gene Details

Gene
C7orf61
Gene Name
chromosome 7 open reading frame 61
There are no transcripts associated with this gene.

Gene
TSC22D4
Gene Name
TSC22 domain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303043.1 2332 UTR 3 NP_001289972.1
NM_030935.4 2332 UTR 3 NP_112197.1

View Full Product Details