Product Details

SNP ID
rs7172522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:88636788 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTCAAGCAAATCCGCCTCAGCCT[A/G]GAGTAACTGGGATTATAGACACTAG
Phenotype
MIM: 610177 MIM: 604533
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AEN PubMed Links

Gene Details

Gene
AEN
Gene Name
apoptosis enhancing nuclease
There are no transcripts associated with this gene.

Gene
ISG20
Gene Name
interferon stimulated exonuclease gene 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303233.1 Intron NP_001290162.1
NM_001303234.1 Intron NP_001290163.1
NM_001303235.1 Intron NP_001290164.1
NM_001303236.1 Intron NP_001290165.1
NM_001303237.1 Intron NP_001290166.1
NM_002201.5 Intron NP_002192.2
XM_005254899.2 Intron XP_005254956.1
XM_006720488.3 Intron XP_006720551.1
XM_011521521.2 Intron XP_011519823.1
XM_017022147.1 Intron XP_016877636.1
XM_017022148.1 Intron XP_016877637.1

View Full Product Details