Product Details
- SNP ID
-
rs3761
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:24951703 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAGTGGTATTTACTTGAGGCAACA[A/G]AATTACGCTTAACAACACACTAAAT
- Phenotype
-
MIM: 162280
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR6841
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79736124] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR6841
- Gene Name
- microRNA 6841
There are no transcripts associated with this gene.
- Gene
- NEFL
- Gene Name
- neurofilament, light polypeptide
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006158.4 |
3093 |
UTR 3 |
|
|
NP_006149.2 |
View Full Product Details