Product Details

SNP ID
hCV12115100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:41531188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATTCTTTCCATTTTCTTCGTCT[C/T]TCTTTGGAGGTCTGGTTGTATCTGC
Phenotype
MIM: 300172
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASK PubMed Links

Gene Details

Gene
CASK
Gene Name
calcium/calmodulin dependent serine protein kinase
There are no transcripts associated with this gene.

View Full Product Details