Product Details

SNP ID
rs61752458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:77902650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCGTTTTCCTGTAAGTTCGAAG[C/A]ATTCAGCAAACAATGGCAAGGCAGA
Phenotype
MIM: 300011 MIM: 300885 MIM: 300715
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
ATP7A PubMed Links

Gene Details

Gene
ATP7A
Gene Name
ATPase copper transporting alpha
There are no transcripts associated with this gene.

Gene
COX7B
Gene Name
cytochrome c oxidase subunit 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001866.2 Intron NP_001857.1
Gene
MAGT1
Gene Name
magnesium transporter 1
There are no transcripts associated with this gene.

View Full Product Details