Product Details

SNP ID
rs5190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:116172778 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAAGCATCTTATATAGTTCCCCT[T/C]GTTTGGTGTATGGCCTGTTTGTCCT
Phenotype
MIM: 300034
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
AGTR2 PubMed Links

Gene Details

Gene
AGTR2
Gene Name
angiotensin II receptor type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000686.4 603 Silent Mutation CTC,CTT L166L NP_000677.2
XM_011537533.1 603 Silent Mutation CTC,CTT L166L XP_011535835.1

View Full Product Details