Product Details
- SNP ID
-
rs10863088
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:85881965 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATATATAGTTAGTAAAGTTTATAG[A/C]AATTTTTTAAAAGAGCTAGCCTGGT
- Phenotype
-
MIM: 162650
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NTS
PubMed Links
Gene Details
- Gene
- NTS
- Gene Name
- neurotensin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006183.4 |
|
Intron |
|
|
NP_006174.1 |
View Full Product Details