Product Details

SNP ID
rs4925133
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:17981487 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGGCAGCAGGGATGACGCTGTAGCC[G/A]TGCAAGCTGCATGCATAGTTGCCCT
Phenotype
MIM: 615519
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
DRC3 PubMed Links

Gene Details

Gene
DRC3
Gene Name
dynein regulatory complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130090.1 Intron NP_001123562.1
NM_001130091.1 Intron NP_001123563.1
NM_001130092.1 Intron NP_001123564.1
NM_031294.3 Intron NP_112584.3
XM_006721583.3 Intron XP_006721646.1
XM_011524018.2 Intron XP_011522320.1
XM_011524019.2 Intron XP_011522321.1
XM_011524020.2 Intron XP_011522322.1
XM_011524021.2 Intron XP_011522323.1
XM_011524022.2 Intron XP_011522324.1
XM_011524023.2 Intron XP_011522325.1
XM_011524025.2 Intron XP_011522327.1
XM_011524026.2 Intron XP_011522328.1
Gene
TOM1L2
Gene Name
target of myb1 like 2 membrane trafficking protein
There are no transcripts associated with this gene.

View Full Product Details