Product Details

SNP ID
rs2000404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:79349952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCCAACCCGCTGCCACTCCAGG[C/T]CAGTGGCAGGCCCTGCCCGGGGGAC
Phenotype
MIM: 604486
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPIF PubMed Links

Gene Details

Gene
PPIF
Gene Name
peptidylprolyl isomerase F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005729.3 Intron NP_005720.1
XM_005269379.2 Intron XP_005269436.1

View Full Product Details