Product Details

SNP ID
rs1418387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92690255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCCGCCGCCGCGCTGGCCGCTG[C/T]CTACGGACCCGGCGGCTTCGGGGGC
Phenotype
MIM: 604420
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HHEX PubMed Links

Gene Details

Gene
HHEX
Gene Name
hematopoietically expressed homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002729.4 332 Missense Mutation GCC,GTC A90V NP_002720.1

View Full Product Details