Product Details

SNP ID
rs11569301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46118303 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAGGGGAGTCAGCAGAGGCCTCG[C/T]TCGGGCGCCCAGTGGTCCTGCCGCC
Phenotype
MIM: 109535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD40 PubMed Links

Gene Details

Gene
CD40
Gene Name
CD40 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001250.5 50 UTR 5 NP_001241.1
NM_001302753.1 50 UTR 5 NP_001289682.1
NM_001322421.1 50 UTR 5 NP_001309350.1
NM_001322422.1 50 UTR 5 NP_001309351.1
NM_152854.3 50 UTR 5 NP_690593.1
XM_005260619.3 50 UTR 5 XP_005260676.1
XM_011529109.2 50 UTR 5 XP_011527411.1
XM_017028135.1 50 UTR 5 XP_016883624.1
XM_017028136.1 50 UTR 5 XP_016883625.1
XM_017028137.1 50 UTR 5 XP_016883626.1

View Full Product Details