Product Details
- SNP ID
-
rs2301363
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:137139979 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GCCACCTTGGCTGGTCCTCAGAGGG[T/C]CCCTGGGGCTCCAGGCCCTGACTGG
- Phenotype
-
MIM: 138249
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GRIN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76135674] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GRIN1
- Gene Name
- glutamate ionotropic receptor NMDA type subunit 1
View Full Product Details