Product Details

SNP ID
rs3006240
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:212731133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTCTCTAGAGAGATATTTTTTT[C/T]TTCAGAGACTTTCATAATATAATCT
Phenotype
MIM: 609174
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NSL1 PubMed Links
Additional Information
For this assay, SNP(s) [rs112737280,rs113022483] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
NSL1
Gene Name
NSL1, MIS12 kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042549.1 8155 UTR 3 NP_001036014.1
NM_001297736.1 8155 UTR 3 NP_001284665.1
NM_001297737.1 8155 UTR 3 NP_001284666.1
NM_001297739.1 8155 Intron NP_001284668.1
NM_015471.3 8155 UTR 3 NP_056286.3
XM_017000968.1 8155 Intron XP_016856457.1
XM_017000969.1 8155 Intron XP_016856458.1

View Full Product Details