Product Details

SNP ID
rs3018301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72081025 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCACCGACCCTCATCACCTTTCAC[A/G]GCGAACAAGATACAACACTCACATG
Phenotype
MIM: 612414 MIM: 164009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRTOMT PubMed Links

Gene Details

Gene
LRTOMT
Gene Name
leucine rich transmembrane and O-methyltransferase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145307.4 Intron NP_001138779.1
NM_001145308.4 Intron NP_001138780.1
NM_001145309.3 Intron NP_001138781.1
NM_001145310.3 Intron NP_001138782.1
NM_001205138.3 Intron NP_001192067.1
NM_001271471.2 Intron NP_001258400.1
NM_001318803.1 Intron NP_001305732.1
NM_145309.5 Intron NP_660352.1
XM_006718473.3 Intron XP_006718536.1
XM_006718474.3 Intron XP_006718537.1
XM_011544847.2 Intron XP_011543149.1
XM_011544848.2 Intron XP_011543150.1
XM_017017356.1 Intron XP_016872845.1
XM_017017357.1 Intron XP_016872846.1
XM_017017358.1 Intron XP_016872847.1
XM_017017359.1 Intron XP_016872848.1
XM_017017360.1 Intron XP_016872849.1
Gene
MIR3165
Gene Name
microRNA 3165
There are no transcripts associated with this gene.

Gene
NUMA1
Gene Name
nuclear mitotic apparatus protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286561.1 Intron NP_001273490.1
NM_006185.3 Intron NP_006176.2
XM_006718564.1 Intron XP_006718627.1
XM_011545054.1 Intron XP_011543356.1
XM_011545055.1 Intron XP_011543357.1
XM_011545056.2 Intron XP_011543358.1
XM_011545057.2 Intron XP_011543359.1
XM_011545058.1 Intron XP_011543360.1
XM_011545059.2 Intron XP_011543361.1
XM_011545060.2 Intron XP_011543362.1
XM_011545061.2 Intron XP_011543363.1
XM_011545062.2 Intron XP_011543364.1
XM_011545063.2 Intron XP_011543365.1
XM_011545064.1 Intron XP_011543366.1
XM_011545065.1 Intron XP_011543367.1
XM_011545066.1 Intron XP_011543368.1
XM_017017830.1 Intron XP_016873319.1
XM_017017831.1 Intron XP_016873320.1

View Full Product Details