Product Details

SNP ID
rs3018385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118257666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGTTGACCTAAGTCATCTTTAT[G/T]GGTTCTATTCAAACTAGAACCTCAG
Phenotype
MIM: 604873 MIM: 611707
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MPZL2 PubMed Links

Gene Details

Gene
MPZL2
Gene Name
myelin protein zero like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005797.3 Intron NP_005788.1
NM_144765.2 Intron NP_658911.1
Gene
MPZL3
Gene Name
myelin protein zero like 3
There are no transcripts associated with this gene.

View Full Product Details