Product Details

SNP ID
rs3018464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112167470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGCGCCTGTGAGAATGTGACGG[A/G]CTGTGACACAAGGAAGGGTGCGAGG
Phenotype
MIM: 611740 MIM: 600953 MIM: 605791
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCO2 PubMed Links

Gene Details

Gene
BCO2
Gene Name
beta-carotene oxygenase 2
There are no transcripts associated with this gene.

Gene
IL18
Gene Name
interleukin 18
There are no transcripts associated with this gene.

Gene
TEX12
Gene Name
testis expressed 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031275.4 99 UTR 5 NP_112565.1

View Full Product Details