Product Details

SNP ID
rs1009457
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:101431023 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAGTTCTCAGCTCTAGGCAGCAGCA[A/T]CTTGGAAGGCTTCTCTGAAGGTGGA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARMCX4 PubMed Links

Gene Details

Gene
ARMCX4
Gene Name
armadillo repeat containing, X-linked 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256155.2 Intron NP_001243084.2

View Full Product Details