Product Details

SNP ID
rs2281307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101430328 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGAAGTTTCGGTGGAAAGAACAC[C/T]GATATTTGTTGAATGATTGCTCTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARMCX4 PubMed Links

Gene Details

Gene
ARMCX4
Gene Name
armadillo repeat containing, X-linked 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256155.2 Intron NP_001243084.2

View Full Product Details