Product Details

SNP ID
rs3027580
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101408672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTGGTCTATCCATTCAGGCACT[C/G]AGTCTCCACTGCCTGATTGTCTGTA
Phenotype
MIM: 300644 MIM: 300610
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ARMCX4 PubMed Links

Gene Details

Gene
ARMCX4
Gene Name
armadillo repeat containing, X-linked 4
There are no transcripts associated with this gene.

Gene
GLA
Gene Name
galactosidase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000169.2 Intron NP_000160.1
Gene
HNRNPH2
Gene Name
heterogeneous nuclear ribonucleoprotein H2 (H')
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032393.2 Intron NP_001027565.1
NM_019597.4 Intron NP_062543.1
Gene
RPL36A-HNRNPH2
Gene Name
RPL36A-HNRNPH2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199973.1 Intron NP_001186902.1
NM_001199974.1 Intron NP_001186903.1

View Full Product Details