Product Details
- SNP ID
-
rs2310883
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:43667819 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATCCTTTCAGATATTTACTGTGCAA[C/G]TATGAAGTATATACATTTTTTGCTA
- Phenotype
-
MIM: 309850
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MAOA
PubMed Links
Gene Details
- Gene
- MAOA
- Gene Name
- monoamine oxidase A
View Full Product Details