Product Details

SNP ID
rs2310958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:406497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCAGGGCACCCTCGCCTGGCCCC[G/A]CTCCCCACCGGAGCCTGGGAAGCTG
Phenotype
MIM: 610336
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
C2CD4C PubMed Links

Gene Details

Gene
C2CD4C
Gene Name
C2 calcium dependent domain containing 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136263.1 1481 UTR 3 NP_001129735.1
XM_011527694.1 1481 Missense Mutation CGG,TGG R440W XP_011525996.1

View Full Product Details