Product Details

SNP ID
rs3027855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136532233 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCAAGCACTAACCTGCTCTGAAGT[A/G]AGCCAGGCAGCTCTGGCCATCTTTT
Phenotype
MIM: 300583
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VGLL1 PubMed Links

Gene Details

Gene
VGLL1
Gene Name
vestigial like family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016267.3 82 UTR 5 NP_057351.1

View Full Product Details