Product Details

SNP ID
rs3027864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136541780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCAAGTCCCAAACACAGAGACC[C/T]GTGTGGCTAGGAAATGCCGTCAAAT
Phenotype
MIM: 300583
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR934 PubMed Links

Gene Details

Gene
MIR934
Gene Name
microRNA 934
There are no transcripts associated with this gene.

Gene
VGLL1
Gene Name
vestigial like family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016267.3 Intron NP_057351.1

View Full Product Details