Product Details

SNP ID
rs2247392
Assay Type
Functionally tested
NCBI dbSNP Submissions
56
Location
Chr.1:6198087 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TTTATGAACCACAATAAAATGAACA[C/T]TTCCTTTTGTTTGCCCTAACATTCC
Phenotype
MIM: 616923 MIM: 180474
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724450 PubMed Links

Gene Details

Gene
LOC102724450
Gene Name
uncharacterized LOC102724450
There are no transcripts associated with this gene.

Gene
RNF207
Gene Name
ring finger protein 207
There are no transcripts associated with this gene.

Gene
RPL22
Gene Name
ribosomal protein L22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000983.3 Intron NP_000974.1

View Full Product Details