Product Details

SNP ID
rs3027917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154022619 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGTCACAGTGTTGAGCCCTGGAG[A/G]ATGGGGTGGGATGTGACGGAAGTGT
Phenotype
MIM: 300283 MIM: 300005 MIM: 300929
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRAK1 PubMed Links

Gene Details

Gene
IRAK1
Gene Name
interleukin 1 receptor associated kinase 1
There are no transcripts associated with this gene.

Gene
MECP2
Gene Name
methyl-CpG binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110792.1 9322 Intron NP_001104262.1
NM_001316337.1 9322 Intron NP_001303266.1
NM_004992.3 9322 UTR 3 NP_004983.1
XM_005274682.4 9322 UTR 3 XP_005274739.1
XM_006724819.3 9322 UTR 3 XP_006724882.1
XM_011531166.2 9322 UTR 3 XP_011529468.1
XM_017029533.1 9322 UTR 3 XP_016885022.1
Gene
MIR718
Gene Name
microRNA 718
There are no transcripts associated with this gene.

View Full Product Details