Product Details

SNP ID
rs3026444
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110284827 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATAAGTGTTTAGGGAGTACTTCT[A/G]TAGAATTAATTTGCTTTGGTTTGTT
Phenotype
MIM: 108740
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ATP2A2 PubMed Links

Gene Details

Gene
ATP2A2
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001681.3 Intron NP_001672.1
NM_170665.3 Intron NP_733765.1
XM_005253888.2 Intron XP_005253945.1
XM_011538402.2 Intron XP_011536704.1

View Full Product Details