Product Details
- SNP ID
-
rs2338200
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
46
- Location
-
Chr.1:151873093 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAAACCACCCTGTGGCTGGAGCC[A/G]AGATACGCTGGCGGCAATGCTGCTG
- Phenotype
-
MIM: 606388
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
THEM4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78345195] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- THEM4
- Gene Name
- thioesterase superfamily member 4
View Full Product Details