Product Details

SNP ID
rs2360672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101224457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAGGCTTCCTTTCTGCGCATGC[A/G]CACAGTAGATTTTTTAAAAATGTTT
Phenotype
MIM: 300052
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DRP2 PubMed Links

Gene Details

Gene
DRP2
Gene Name
dystrophin related protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171184.1 Intron NP_001164655.1
NM_001939.2 Intron NP_001930.2
XM_017029333.1 Intron XP_016884822.1
XM_017029334.1 Intron XP_016884823.1

View Full Product Details